Order Entry
Ireland
ContactUsLinkComponent
 
Recombinant Human CCL11 (from E. coli)
Catalog # ABNOP7211.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CCL11 (from E. coli)
Catalog # ABNOP7211.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    P51671
  • Protein/peptide name:
    CCL11
  • Sequence:
    24a.a.-97a.a.

Specifications

About this item