Recombinant Human CCL21 (from E. coli)
Catalog # ABNOP7219.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:O00585
- Protein/peptide name:CCL21
- Sequence:24a.a.-134a.a.