Recombinant Human IL1RN (from E. coli)
Catalog # ABNOP7127.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P81510-1
- Protein/peptide name:IL1RN
- Sequence:26a.a.-177a.a.