Recombinant Human LIF (from E. coli)
Catalog # ABNOP7142.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P15018
- Protein/peptide name:LIF
- Sequence:23a.a.-202a.a.