Order Entry
Ireland
ContactUsLinkComponent
Anti-Cd86 Rabbit Recombinant Antibody [clone: PO.3]
Anti-Cd86 Rabbit Recombinant Antibody [clone: PO.3]
Catalog # ABNORAB03348.200UG
Supplier:  Abnova
Anti-Cd86 Rabbit Recombinant Antibody [clone: PO.3]
Catalog # ABNORAB03348.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Cd86
  • Clonality:
    Recombinant
  • Clone:
    PO.3
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against mouse Cd86.
  • Pk:
    200 µG

Specifications

About this item