Anti-DCLK1 Rabbit Recombinant Antibody [clone: R05-8D6]
ABNORAB02330.100UL
: Abnova
- Antigen symbol:DCLK1
- Clonality:Recombinant
- Clone:R05-8D6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human DCLK1.
- Pk:100 µl