Anti-DBF4 Rabbit Recombinant Antibody [clone: R06-6I2]
Catalog # ABNORAB02331.100UL
Supplier: Abnova
Specifications
- Antigen symbol:DBF4
- Clonality:Recombinant
- Clone:R06-6I2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human DBF4.
- Pk:100 µl