Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD11b Rabbit Recombinant Antibody [clone: R01-3F9]
Catalog # ABNORAB01664.100UL
Supplier:  Abnova
Anti-CD11b Rabbit Recombinant Antibody [clone: R01-3F9]
Catalog # ABNORAB01664.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CD11b
  • Clonality:
    Recombinant
  • Clone:
    R01-3F9
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of mouse CD11b
  • Pk:
    100 µl

Specifications

About this item