Anti-CD8 Rabbit Recombinant Antibody [clone: R09-8E7]
Catalog # ABNORAB01665.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CD8
- Clonality:Recombinant
- Clone:R09-8E7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CD8
- Pk:100 µl