Anti-Src (pTyr419) Rabbit Recombinant Antibody [clone: R01-6H3] ()
Catalog # ABNORAB01672.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Src
- Clonality:Recombinant
- Clone:R01-6H3
- Host:Rabbit
- Reactivity:Human
- Modification:pTyr419
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic phosphopeptide corresponding to residues surrounding Tyr419 of human Src
- Pk:100 µl