Anti-Pirh2 Rabbit Recombinant Antibody [clone: R06-8H6]
ABNORAB01712.100UL
: Abnova
- Antigen symbol:Pirh2
- Clonality:Recombinant
- Clone:R06-8H6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Pirh2
- Pk:100 µl