Anti-TREX1 Rabbit Recombinant Antibody [clone: R05-1D6]
Catalog # ABNORAB01714.100UL
Supplier: Abnova
Specifications
- Antigen symbol:TREX1
- Clonality:Recombinant
- Clone:R05-1D6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TREX1
- Pk:100 µl