Order Entry
Ireland
ContactUsLinkComponent
Anti-RAB5C Rabbit Recombinant Antibody [clone: R04-3B7]
Anti-RAB5C Rabbit Recombinant Antibody [clone: R04-3B7]
Catalog # ABNORAB01716.100UL
Supplier:  Abnova
Anti-RAB5C Rabbit Recombinant Antibody [clone: R04-3B7]
Catalog # ABNORAB01716.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    RAB5C
  • Clonality:
    Recombinant
  • Clone:
    R04-3B7
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human RAB5C
  • Pk:
    100 µl

Specifications

About this item