Anti-DNA Polymerase delta, catalytic subunit Rabbit Recombinant Antibody [clone: R05-3H1]
Catalog # ABNORAB01717.100UL
Supplier: Abnova
Specifications
- Antigen symbol:DNA Polymerase delta, catalytic subunit
- Clonality:Recombinant
- Clone:R05-3H1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human DNA Polymerase delta, catalytic subunit
- Pk:100 µl