Anti-Desmocollin 3 Rabbit Recombinant Antibody [clone: R06-4G2]
ABNORAB01719.100UL
: Abnova
- Antigen symbol:Desmocollin 3
- Clonality:Recombinant
- Clone:R06-4G2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Desmocollin 3
- Pk:100 µl