Order Entry
Ireland
ContactUsLinkComponent
Anti-Menin Rabbit Recombinant Antibody [clone: R05-6A9]
Anti-Menin Rabbit Recombinant Antibody [clone: R05-6A9]
Catalog # ABNORAB01723.100UL
Supplier:  Abnova
Anti-Menin Rabbit Recombinant Antibody [clone: R05-6A9]
Catalog # ABNORAB01723.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Menin
  • Clonality:
    Recombinant
  • Clone:
    R05-6A9
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Menin
  • Pk:
    100 µl

Specifications

About this item