Anti-CENPA Rabbit Recombinant Antibody [clone: R04-9A5]
Catalog # ABNORAB01727.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CENPA
- Clonality:Recombinant
- Clone:R04-9A5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CENPA
- Pk:100 µl