Anti-TNFAIP8 Rabbit Recombinant Antibody [clone: R04-5F3]
Catalog # ABNORAB01739.100UL
Supplier: Abnova
Specifications
- Antigen symbol:TNFAIP8
- Clonality:Recombinant
- Clone:R04-5F3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human TNFAIP8
- Pk:100 µl