Anti-NRF1 Rabbit Recombinant Antibody [clone: R02-9A7]
Catalog # ABNORAB02049.100UL
Supplier: Abnova
Specifications
- Antigen symbol:NRF1
- Clonality:Recombinant
- Clone:R02-9A7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human NRF1.
- Pk:100 µl