Order Entry
Ireland
ContactUsLinkComponent
Anti-NQO1 Rabbit Recombinant Antibody [clone: R02-4D6]
Anti-NQO1 Rabbit Recombinant Antibody [clone: R02-4D6]
Catalog # ABNORAB02051.100UL
Supplier:  Abnova
Anti-NQO1 Rabbit Recombinant Antibody [clone: R02-4D6]
Catalog # ABNORAB02051.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    NQO1
  • Clonality:
    Recombinant
  • Clone:
    R02-4D6
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human NQO1.
  • Pk:
    100 µl

Specifications

About this item