Anti-SLC12A1 Rabbit Recombinant Antibody [clone: R07-5D1]
Catalog # ABNORAB02058.100UL
Supplier: Abnova
Specifications
- Antigen symbol:SLC12A1
- Clonality:Recombinant
- Clone:R07-5D1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SLC12A1.
- Pk:100 µl