Anti-OLIG1 Rabbit Recombinant Antibody [clone: R07-1H5]
Catalog # ABNORAB01395.100UL
Supplier: Abnova
Specifications
- Antigen symbol:OLIG1
- Clonality:Recombinant
- Clone:R07-1H5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human OLIG1.
- Pk:100 µl