Anti-TF Rabbit Recombinant Antibody [clone: R03-4K6]
ABNORAB01856.100UL
: Abnova
- Antigen symbol:TF
- Clonality:Recombinant
- Clone:R03-4K6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TF.
- Pk:100 µl