Anti-Cortactin Rabbit Recombinant Antibody [clone: R07-2C5]
Catalog # ABNORAB01495.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Cortactin
- Clonality:Recombinant
- Clone:R07-2C5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Cortactin
- Pk:100 µl