Anti-cIAP1 Rabbit Recombinant Antibody [clone: R01-1C5]
ABNORAB01501.100UL
: Abnova
- Antigen symbol:cIAP1
- Clonality:Recombinant
- Clone:R01-1C5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human cIAP1.
- Pk:100 µl