Anti-Chk2 Rabbit Recombinant Antibody [clone: R04-8E5]
Catalog # ABNORAB01503.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Chk2
- Clonality:Recombinant
- Clone:R04-8E5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Chk2.
- Pk:100 µl