Order Entry
Ireland
ContactUsLinkComponent
Anti-Cdk4 Rabbit Recombinant Antibody [clone: R07-4G7]
Anti-Cdk4 Rabbit Recombinant Antibody [clone: R07-4G7]
Catalog # ABNORAB01513.100UL
Supplier:  Abnova
Anti-Cdk4 Rabbit Recombinant Antibody [clone: R07-4G7]
Catalog # ABNORAB01513.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Cdk4
  • Clonality:
    Recombinant
  • Clone:
    R07-4G7
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Cdk4.
  • Pk:
    100 µl

Specifications

About this item