Order Entry
Ireland
ContactUsLinkComponent
Anti-CDC7 Rabbit Recombinant Antibody [clone: R01-4G6]
Anti-CDC7 Rabbit Recombinant Antibody [clone: R01-4G6]
Catalog # ABNORAB01517.100UL
Supplier:  Abnova
Anti-CDC7 Rabbit Recombinant Antibody [clone: R01-4G6]
Catalog # ABNORAB01517.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CDC7
  • Clonality:
    Recombinant
  • Clone:
    R01-4G6
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human CDC7.
  • Pk:
    100 µl

Specifications

About this item