Anti-Bak Rabbit Recombinant Antibody [clone: R03-6A8]
Catalog # ABNORAB01566.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Bak
- Clonality:Recombinant
- Clone:R03-6A8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Bak.
- Pk:100 µl