Order Entry
Ireland
ContactUsLinkComponent
Anti-Catalase Rabbit Recombinant Antibody [clone: R01-8B2]
Anti-Catalase Rabbit Recombinant Antibody [clone: R01-8B2]
Catalog # ABNORAB01534.100UL
Supplier:  Abnova
Anti-Catalase Rabbit Recombinant Antibody [clone: R01-8B2]
Catalog # ABNORAB01534.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Catalase
  • Clonality:
    Recombinant
  • Clone:
    R01-8B2
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Catalase.
  • Pk:
    100 µl

Specifications

About this item