Anti-Catalase Rabbit Recombinant Antibody [clone: R03-8E9]
Catalog # ABNORAB01535.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Catalase
- Clonality:Recombinant
- Clone:R03-8E9
- Host:Rabbit
- Reactivity:Mouse
- Immunogen:Recombinant rabbit monoclonal antibody raised against mouse Catalase.
- Pk:100 µl