Anti-BMP2 Rabbit Recombinant Antibody [clone: R03-3H9]
Catalog # ABNORAB01554.100UL
Supplier: Abnova
Specifications
- Antigen symbol:BMP2
- Clonality:Recombinant
- Clone:R03-3H9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human BMP2
- Pk:100 µl