Order Entry
Ireland
ContactUsLinkComponent
Anti-Bsg Rabbit Recombinant Antibody [clone: R03-7C3]
Anti-Bsg Rabbit Recombinant Antibody [clone: R03-7C3]
Catalog # ABNORAB01314.100UL
Supplier:  Abnova
Anti-Bsg Rabbit Recombinant Antibody [clone: R03-7C3]
Catalog # ABNORAB01314.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Bsg
  • Clonality:
    Recombinant
  • Clone:
    R03-7C3
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against mouse Bsg.
  • Pk:
    100 µl

Specifications

About this item