Anti-H3C1 Rabbit Recombinant Antibody [clone: R08-9D6]
Catalog # ABNORAB01317.100UL
Supplier: Abnova
Specifications
- Antigen symbol:H3C1
- Clonality:Recombinant
- Clone:R08-9D6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human H3C1.
- Pk:100 µl