Anti-MALE Rabbit Recombinant Antibody [clone: R02-6F4]
ABNORAB01319.100UL
: Abnova
- Antigen symbol:MALE
- Clonality:Recombinant
- Clone:R02-6F4
- Host:Rabbit
- Reactivity:E. coli
- Immunogen:Recombinant rabbit monoclonal antibody raised against Escherichia coli malE.
- Pk:100 µl