Anti-CCNA2/CCNA1 Rabbit Recombinant Antibody [clone: R07-3D4]
Catalog # ABNORAB01320.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CCNA2/CCNA1
- Clonality:Recombinant
- Clone:R07-3D4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human CCNA2/CCNA1.
- Pk:100 µl