Anti-AKT1 Rabbit Recombinant Antibody [clone: R07-4A7]
Catalog # ABNORAB01796.100UL
Supplier: Abnova
Specifications
- Antigen symbol:AKT1
- Clonality:Recombinant
- Clone:R07-4A7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human AKT1.
- Pk:100 µl