Order Entry
Ireland
ContactUsLinkComponent
Anti-PIM1 Rabbit Recombinant Antibody [clone: R08-8D5]
Anti-PIM1 Rabbit Recombinant Antibody [clone: R08-8D5]
Catalog # ABNORAB01804.100UL
Supplier:  Abnova
Anti-PIM1 Rabbit Recombinant Antibody [clone: R08-8D5]
Catalog # ABNORAB01804.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    PIM1
  • Clonality:
    Recombinant
  • Clone:
    R08-8D5
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PIM1.
  • Pk:
    100 µl

Specifications

About this item