Order Entry
Ireland
ContactUsLinkComponent
 
Anti-PAK1 Rabbit Recombinant Antibody [clone: R02-5G3]
  ABNORAB01807.100UL
 :  Abnova
Anti-PAK1 Rabbit Recombinant Antibody [clone: R02-5G3]
  ABNORAB01807.100UL
 :  Abnova

 

  • Antigen symbol:
    PAK1
  • Clonality:
    Recombinant
  • Clone:
    R02-5G3
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PAK1.
  • Pk:
    100 µl