Order Entry
Ireland
ContactUsLinkComponent
Anti-IRF7 Rabbit Recombinant Antibody [clone: R04-1E1]
Anti-IRF7 Rabbit Recombinant Antibody [clone: R04-1E1]
Catalog # ABNORAB01814.100UL
Supplier:  Abnova
Anti-IRF7 Rabbit Recombinant Antibody [clone: R04-1E1]
Catalog # ABNORAB01814.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    IRF7
  • Clonality:
    Recombinant
  • Clone:
    R04-1E1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human IRF7.
  • Pk:
    100 µl

Specifications

About this item