Order Entry
Ireland
ContactUsLinkComponent
Anti-YBX1 Rabbit Recombinant Antibody [clone: R03-6G4]
Anti-YBX1 Rabbit Recombinant Antibody [clone: R03-6G4]
  ABNORAB01817.100UL
 :  Abnova
Anti-YBX1 Rabbit Recombinant Antibody [clone: R03-6G4]
  ABNORAB01817.100UL
 :  Abnova

 

  • Antigen symbol:
    YBX1
  • Clonality:
    Recombinant
  • Clone:
    R03-6G4
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human YBX1.
  • Pk:
    100 µl