Anti-NHEJ1 Rabbit Recombinant Antibody [clone: R08-9D7]
Catalog # ABNORAB01822.100UL
Supplier: Abnova
Specifications
- Antigen symbol:NHEJ1
- Clonality:Recombinant
- Clone:R08-9D7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human NHEJ1.
- Pk:100 µl