Anti-ACTA2 Rabbit Recombinant Antibody [clone: R01-8C7]
Catalog # ABNORAB01299.100UL
Supplier: Abnova
Specifications
- Antigen symbol:ACTA2
- Clonality:Recombinant
- Clone:R01-8C7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ACTA2.
- Pk:100 µl