Anti-ACTL6A Rabbit Recombinant Antibody [clone: R03-1G4]
ABNORAB01768.100UL
: Abnova
- Antigen symbol:ACTL6A
- Clonality:Recombinant
- Clone:R03-1G4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ACTL6A.
- Pk:100 µl