Order Entry
Ireland
ContactUsLinkComponent
Anti-PCNA3 Rabbit Recombinant Antibody [clone: R09-6B3]
Anti-PCNA3 Rabbit Recombinant Antibody [clone: R09-6B3]
  ABNORAB01235.100UL
 :  Abnova
Anti-PCNA3 Rabbit Recombinant Antibody [clone: R09-6B3]
  ABNORAB01235.100UL
 :  Abnova

 

  • Antigen symbol:
    PCNA3
  • Clonality:
    Recombinant
  • Clone:
    R09-6B3
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PCNA3.
  • Pk:
    100 µl