Anti-UBE2I Rabbit Recombinant Antibody [clone: R01-6I6]
Catalog # ABNORAB01207.100UL
Supplier: Abnova
Specifications
- Antigen symbol:UBE2I
- Clonality:Recombinant
- Clone:R01-6I6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human UBE2I.
- Pk:100 µl