Anti-SMARCA2 Rabbit Recombinant Antibody [clone: R02-3E2]
Catalog # ABNORAB01212.100UL
Supplier: Abnova
Specifications
- Antigen symbol:SMARCA2
- Clonality:Recombinant
- Clone:R02-3E2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SMARCA2.
- Pk:100 µl