Anti-PTK2 Rabbit Recombinant Antibody [clone: R05-1A1]
Catalog # ABNORAB01227.100UL
Supplier: Abnova
Specifications
- Antigen symbol:PTK2
- Clonality:Recombinant
- Clone:R05-1A1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PTK2.
- Pk:100 µl