Anti-Ythdf1 Rabbit Recombinant Antibody [clone: R05-2E7]
Catalog # ABNORAB01193.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Ythdf1
- Clonality:Recombinant
- Clone:R05-2E7
- Host:Rabbit
- Reactivity:Mouse
- Immunogen:Recombinant rabbit monoclonal antibody raised against mouse Ythdf1.
- Pk:100 µl