Order Entry
Ireland
ContactUsLinkComponent
Anti-FLT1 Rabbit Recombinant Antibody [clone: R06-1A4]
Anti-FLT1 Rabbit Recombinant Antibody [clone: R06-1A4]
Catalog # ABNORAB01198.100UL
Supplier:  Abnova
Anti-FLT1 Rabbit Recombinant Antibody [clone: R06-1A4]
Catalog # ABNORAB01198.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    FLT1
  • Clonality:
    Recombinant
  • Clone:
    R06-1A4
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human FLT1.
  • Pk:
    100 µl

Specifications

About this item